Gene Transcripts

Gene Symbol
Ctsr
Entrez Gene ID
290975
Gene Name
cathepsin R
Gene Aliases
Cts6
Chromosome Location
Chr. 17: 3885684 - 3894447 on Build Rnor_6.0
UniGene ID
Rn.106060
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175581.3 NP_783171.2 6 735
XM_008771434.1 XP_008769656.1 5 656
GenBank mRNA AF456459.1 - 695
BC097484.1 - 6 735 7459242

View Full Product Details