Gene Transcripts
- Gene Symbol
- ANKRD26P1
- Entrez Gene ID
- 124149
- Gene Name
- ankyrin repeat domain 26 pseudogene 1
- Gene Aliases
- Chromosome Location
- Chr. 16: 46469337 - 46569097 on Build GRCh38
- UniGene ID
- Hs.97414
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_026556.1 |
|
12 |
2654 |
|
GenBank mRNA |
BC070117.1 |
- |
12 |
2654 |
|
BX648636.1 |
- |
2 |
437 |
|
View Full Product Details