Gene Transcripts
- Gene Symbol
- Abhd14a
- Entrez Gene ID
- 300982
- Gene Name
- abhydrolase domain containing 14A
- Gene Aliases
- Dorz1, RGD1309721
- Chromosome Location
- Chr. 8: 115141575 - 115151219 on Build Rnor_6.0
- UniGene ID
- Rn.41351
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001009670.1 |
NP_001009670.1
|
4, 5 |
833 |
|
XM_006243717.2 |
XP_006243779.1
|
3, 4 |
674 |
|
XM_006243720.2 |
XP_006243782.1
|
3, 4 |
1428 |
|
XM_008766494.1 |
XP_008764716.1
|
4, 5 |
1245 |
|
XM_008766495.1 |
XP_008764717.1
|
5, 6 |
899 |
|
XM_008766496.1 |
XP_008764718.1
|
4, 5 |
848 |
|
XM_008766497.1 |
XP_008764719.1
|
4, 5 |
717 |
|
XM_008766498.1 |
XP_008764720.1
|
3, 4 |
669 |
|
XM_008766499.1 |
XP_008764721.1
|
3, 4 |
674 |
|
GenBank mRNA |
BC088472.1 |
- |
4, 5 |
833 |
|
View Full Product Details