Gene Transcripts

Gene Symbol
Fastkd2
Entrez Gene ID
301463
Gene Name
FAST kinase domains 2
Gene Aliases
RGD1307883
Chromosome Location
Chr. 9: 70448386 - 70471420 on Build Rnor_6.0
UniGene ID
Rn.24905
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001009673.1 NP_001009673.1 1 409
XM_006245050.1 XP_006245112.1 2 536
GenBank mRNA BC088416.1 - 1 409

View Full Product Details