Gene Transcripts

Gene Symbol
OSBPL7
Entrez Gene ID
114881
Gene Name
oxysterol binding protein like 7
Gene Aliases
ORP7
Chromosome Location
Chr. 17: 47807367 - 47821822 on Build GRCh38
UniGene ID
Hs.463320
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145798.2 NP_665741.1 3 365
XM_017024122.1 XP_016879611.1 3 387
XM_017024123.1 XP_016879612.1 3 505
XM_017024124.1 XP_016879613.1 3 406
GenBank mRNA AF323729.1 - 3 354
AK289523.1 - 3 387
BC028010.1 - 3 364
BC065482.1 - 3 337 6081784

View Full Product Details