Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001011966.1 | NP_001011966.1 | 12, 13 | 1390 | |
XM_006235883.2 | XP_006235945.1 | 11, 12 | 1487 | ||
XM_006235884.2 | XP_006235946.1 | 11, 12 | 1487 | ||
XM_006235885.2 | XP_006235947.1 | 11, 12 | 1451 | ||
XM_006235886.2 | XP_006235948.1 | 12, 13 | 1474 | ||
XM_006235887.2 | XP_006235949.1 | 12, 13 | 1438 | ||
XM_006235888.2 | XP_006235950.1 | 11, 12 | 1487 | ||
XM_006235889.2 | XP_006235951.1 | 11, 12 | 1451 | ||
GenBank mRNA | BC085349.1 | - | 12, 13 | 1390 |