Gene Transcripts

Gene Symbol
Morf4l1
Entrez Gene ID
300891
Gene Name
mortality factor 4 like 1
Gene Aliases
Chromosome Location
Chr. 8: 97471523 - 97494974 on Build Rnor_6.0
UniGene ID
Rn.156201
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001011999.1 NP_001011999.1 12 1340
XM_008766445.1 XP_008764667.1 13 1422
GenBank mRNA BC078910.1 - 12 1340
FQ213551.1 - 6 860

View Full Product Details