Gene Transcripts

Gene Symbol
Trim42
Entrez Gene ID
301106
Gene Name
tripartite motif-containing 42
Gene Aliases
Chromosome Location
Chr. 8: 105205040 - 105226580 on Build Rnor_6.0
UniGene ID
Rn.109830
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013955.1 NP_001013977.1 3 1918
XM_008766513.1 XP_008764735.1 3 1963
GenBank mRNA BC087151.1 - 3 1918

View Full Product Details