Gene Transcripts

Gene Symbol
Nif3l1
Entrez Gene ID
301431
Gene Name
NIF3 NGG1 interacting factor 3-like 1 (S. cerevisiae)
Gene Aliases
Chromosome Location
Chr. 9: 65329142 - 65349280 on Build Rnor_6.0
UniGene ID
Rn.229097
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001024763.1 NP_001019934.1 3 735
XM_006244958.2 XP_006245020.1 5 876
XM_006244960.2 XP_006245022.1 5 847
XM_006244961.2 XP_006245023.1 5 872
XM_006244962.2 XP_006245024.1 4 745
XM_006244963.2 XP_006245025.1 5 879
XM_008767082.1 XP_008765304.1 5 880
GenBank mRNA BC097437.1 - 3 735
BC097992.1 - 5 808 7446502

View Full Product Details