Gene Transcripts

Gene Symbol
Prickle3
Entrez Gene ID
317380
Gene Name
prickle homolog 3 (Drosophila)
Gene Aliases
Chromosome Location
Chr. X: 15682652 - 15693473 on Build Rnor_6.0
UniGene ID
Rn.155951
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001014110.1 NP_001014132.1 7 914
GenBank mRNA BC085918.1 - 7 914

View Full Product Details