Gene Transcripts

Gene Symbol
Abhd14a
Entrez Gene ID
300982
Gene Name
abhydrolase domain containing 14A
Gene Aliases
Dorz1, RGD1309721
Chromosome Location
Chr. 8: 115141575 - 115151219 on Build Rnor_6.0
UniGene ID
Rn.41351
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001009670.1 NP_001009670.1 6 1373
XM_006243717.2 XP_006243779.1 5 1214
XM_006243720.2 XP_006243782.1 5 1968
XM_008766494.1 XP_008764716.1 6 1785
XM_008766495.1 XP_008764717.1 7 1439
XM_008766496.1 XP_008764718.1 6 1388
XM_008766497.1 XP_008764719.1 6 1257
XM_008766498.1 XP_008764720.1 5 1209
XM_008766499.1 XP_008764721.1 5 1082
GenBank mRNA BC088472.1 - 6 1373

View Full Product Details