Gene Transcripts

Gene Symbol
NHLRC3
Entrez Gene ID
387921
Gene Name
NHL repeat containing 3
Gene Aliases
Chromosome Location
Chr. 13: 39038311 - 39050109 on Build GRCh38
UniGene ID
Hs.507783
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001012754.3 NP_001012772.1 3 616
NM_001017370.2 NP_001017370.1 3 616
NR_073109.1 3 358
GenBank mRNA AK299198.1 - 3 529
AK300260.1 - 3 529
AL833329.1 - 2 1033
BC052492.1 - 3 296
BC140877.1 - 3 542 9021745
BC140878.1 - 3 542 9021746
BX647872.1 - 3 506
BX648122.1 - 3 568
CR749318.1 - 3 537
DA772703.1 - 3 358

View Full Product Details