Gene Transcripts

Gene Symbol
FRRS1
Entrez Gene ID
391059
Gene Name
ferric chelate reductase 1
Gene Aliases
SDFR2, SDR2
Chromosome Location
Chr. 1: 99708699 - 99766630 on Build GRCh38
UniGene ID
Hs.454779
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013660.2 NP_001013682.2 12, 13 1936
XM_005270861.3 XP_005270918.1 12, 13 1463
XM_011541451.2 XP_011539753.1 12, 13 1936
XM_011541452.1 XP_011539754.1 12, 13 1463
XM_017001271.1 XP_016856760.1 10, 11 1347
GenBank mRNA AK131302.1 - 12, 13 1936

View Full Product Details