Gene Transcripts

Gene Symbol
UBE2R2
Entrez Gene ID
54926
Gene Name
ubiquitin conjugating enzyme E2 R2
Gene Aliases
CDC34B, E2-CDC34B, UBC3B
Chromosome Location
Chr. 9: 33817164 - 33920403 on Build GRCh38
UniGene ID
Hs.643648
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017811.3 NP_060281.2 3 920
XM_005251496.2 XP_005251553.1 2 853
XM_011517949.1 XP_011516251.1 3 940
XM_017014860.1 XP_016870349.1 2 853
GenBank mRNA AJ240087.1 - 3 351
AK000426.1 - 3 537
AK225880.1 - 3 525
BC004862.2 - 3 527 3838157
BC047584.1 - 3 776 4827644
CR457233.1 - 3 346

View Full Product Details