Gene Transcripts

Gene Symbol
Krtap4-16
Entrez Gene ID
435285
Gene Name
keratin associated protein 4-16
Gene Aliases
OTTMUSG00000002196
Chromosome Location
Chr. 11: 99850655 - 99851605 on Build GRCm38
UniGene ID
Mm.351895
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013823.1 NP_001013845.1 1 785
GenBank mRNA BC016249.1 - 1 785

View Full Product Details