Gene Transcripts

Gene Symbol
Tmem5
Entrez Gene ID
299841
Gene Name
transmembrane protein 5
Gene Aliases
Chromosome Location
Chr. 7: 64329341 - 64341196 on Build Rnor_6.0
UniGene ID
Rn.161056
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001024759.1 NP_001019930.1 4 717
XM_006241397.2 XP_006241459.1 4 782
GenBank mRNA BC097355.1 - 4 717

View Full Product Details