Gene Transcripts

Gene Symbol
Adcy1
Entrez Gene ID
432530
Gene Name
adenylate cyclase 1
Gene Aliases
AC1, D11Bwg1392e, I-AC, brl, mKIAA4070
Chromosome Location
Chr. 11: 7063489 - 7178506 on Build GRCm38
UniGene ID
Mm.259733
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009622.1 NP_033752.1 20 7136
GenBank mRNA AK220469.1 - 14 5607
BC019590.1 - 1 1362
BC048830.1 - 1 1362

View Full Product Details