Gene Transcripts

Gene Symbol
Cesl1
Entrez Gene ID
501232
Gene Name
carboxylesterase-like 1
Gene Aliases
Chromosome Location
Chr. : - on Build
UniGene ID
Rn.202952
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001024365.1 NP_001019536.1 9, 10 1061
GenBank mRNA BC079129.1 - 9, 10 1061
U10698.1 - 8, 9 948

View Full Product Details