Gene Transcripts

Gene Symbol
Ankib1
Entrez Gene ID
70797
Gene Name
ankyrin repeat and IBR domain containing 1
Gene Aliases
2310061P20Rik, 4631416I11Rik, AW494740, C80642, mKIAA1386
Chromosome Location
Chr. 5: 3689999 - 3803124 on Build GRCm38
UniGene ID
Mm.130148
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001003909.4 NP_001003909.2 20 3263
NM_001289527.1 NP_001276456.1 20 3281
NM_001289528.1 NP_001276457.1 19 3145
NM_001289529.1 NP_001276458.1 20 2890
GenBank mRNA AK129343.1 - 20 3064
BC079620.1 - 9 953
BC089031.1 - 20 2869
BC145510.1 - 19 2896
BC145511.1 - 20 3032 9054074
BC150792.1 - 20 3032 9087703

View Full Product Details