Gene Transcripts

Gene Symbol
Mrps18c
Entrez Gene ID
68735
Gene Name
mitochondrial ribosomal protein S18C
Gene Aliases
1110037D14Rik
Chromosome Location
Chr. 5: 100798759 - 100804467 on Build GRCm38
UniGene ID
Mm.25585
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026826.1 NP_081102.1 3 233
GenBank mRNA AK004139.1 - 3 246
AK088290.1 - 3 246
AK145935.1 - 3 250
BC028442.1 - 3 233 1196057

View Full Product Details