Gene Transcripts

Gene Symbol
C8orf48
Entrez Gene ID
157773
Gene Name
chromosome 8 open reading frame 48
Gene Aliases
Chromosome Location
Chr. 8: 13566843 - 13568288 on Build GRCh38
UniGene ID
Hs.104941
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001007090.2 NP_001007091.1 1 487
GenBank mRNA AF394908.1 - 1 340
AK058131.1 - 1 487
BC031245.1 - 1 435 5266320

View Full Product Details