Gene Transcripts

Gene Symbol
Clrn3
Entrez Gene ID
309082
Gene Name
clarin 3
Gene Aliases
RGD1305043, Tmem12
Chromosome Location
Chr. 1: 207750627 - 207766250 on Build Rnor_6.0
UniGene ID
Rn.14775
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001014026.1 NP_001014048.1 654
GenBank mRNA BC078942.1 - 654

View Full Product Details