Gene Transcripts

Gene Symbol
Kdm6b
Entrez Gene ID
216850
Gene Name
KDM1 lysine (K)-specific demethylase 6B
Gene Aliases
1700064E03Rik, BC038313, Jmjd3, mKIAA0346
Chromosome Location
Chr. 11: 69398508 - 69418148 on Build GRCm38
UniGene ID
Mm.261201
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001017426.1 NP_001017426.1 19 4741
GenBank mRNA AK129121.1 - 9 1533
AK136019.1 - 9 922
BC038313.1 - 4 379
BC075632.1 - 20 4917 30362899

View Full Product Details