Gene Transcripts

Gene Symbol
Aspscr1
Entrez Gene ID
68938
Gene Name
alveolar soft part sarcoma chromosome region, candidate 1 (human)
Gene Aliases
1190006K01Rik, ASPC, ASPCR1, ASPL, ASPS, RCC17, TUG
Chromosome Location
Chr. 11: 120672439 - 120709447 on Build GRCm38
UniGene ID
Mm.294020
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001164224.1 NP_001157696.1 6 423
NM_026877.2 NP_081153.1 7 644
NM_198223.2 NP_937866.1 6 423
GenBank mRNA AK088197.1 - 7 495
AK146061.1 - 6 424
AY349132.1 - 5 413
AY349133.1 - 7 561
BC019177.1 - 7 587 5008127
BC022115.1 - 7 591 5101238
BC031153.1 - 7 576 4976569

View Full Product Details