Gene Transcripts

Gene Symbol
PLEKHJ1
Entrez Gene ID
55111
Gene Name
pleckstrin homology domain containing J1
Gene Aliases
GNRPX
Chromosome Location
Chr. 19: 2229952 - 2236724 on Build GRCh38
UniGene ID
Hs.501353
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018049.2 NP_060519.1 6 836
XM_017026924.1 XP_016882413.1 4 1339
XM_017026925.1 XP_016882414.1 4 1330
XM_017026926.1 XP_016882415.1 5 1251
XM_017026927.1 XP_016882416.1 5 1242
XM_017026928.1 XP_016882417.1 6 1150
GenBank mRNA AK001159.1 - 6 812
AK092742.1 - 3 2453
AK097723.1 - 7 1843
AK098725.1 - 6 926
BC003084.2 - 6 757 2988404
BC008966.2 - 6 783 2961278

View Full Product Details