Gene Transcripts

Gene Symbol
Efhc2
Entrez Gene ID
74405
Gene Name
EF-hand domain (C-terminal) containing 2
Gene Aliases
4933407D04Rik, mRib72-2
Chromosome Location
Chr. X: 17132049 - 17320461 on Build GRCm38
UniGene ID
Mm.248531
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028916.4 NP_083192.2 8 1256
GenBank mRNA AK016714.1 - 8 1215
AK029501.1 - 8 1222
BC120659.1 - 8 1191 40129582
BC125625.1 - 8 1191 40130140

View Full Product Details