Gene Transcripts

Gene Symbol
Lppr5
Entrez Gene ID
75769
Gene Name
lipid phosphate phosphatase-related protein type 5
Gene Aliases
4833424O15Rik, PRG-5, Pap2d
Chromosome Location
Chr. 3: 117573063 - 117689507 on Build GRCm38
UniGene ID
Mm.72187
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK032034.1 - 6 2245
AK043629.1 - 6 1854
AK076176.1 - 6 1995
AK082692.1 - 6 2142
BC031879.1 - 6 2094 4527789

View Full Product Details