Gene Transcripts
- Gene Symbol
- Lppr5
- Entrez Gene ID
- 75769
- Gene Name
- lipid phosphate phosphatase-related protein type 5
- Gene Aliases
- 4833424O15Rik, PRG-5, Pap2d
- Chromosome Location
- Chr. 3: 117573063 - 117689507 on Build GRCm38
- UniGene ID
- Mm.72187
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK043629.1 |
- |
6 |
3582 |
|
AK076176.1 |
- |
6 |
3722 |
|
View Full Product Details