Gene Transcripts

Gene Symbol
Klhdc10
Entrez Gene ID
76788
Gene Name
kelch domain containing 10
Gene Aliases
2410127E18Rik, AI593221, mKIAA0265
Chromosome Location
Chr. 6: 30401517 - 30455179 on Build GRCm38
UniGene ID
Mm.277308
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029742.2 NP_084018.1 10 4128
GenBank mRNA AK046336.1 - 1 1572
BC056496.1 - 1 1141
BC060132.1 - 10 4128 6409751

View Full Product Details