Gene Transcripts

Gene Symbol
OR10H1
Entrez Gene ID
26539
Gene Name
olfactory receptor family 10 subfamily H member 1
Gene Aliases
Chromosome Location
Chr. 19: 15806957 - 15808759 on Build GRCh38
UniGene ID
Hs.533925
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013940.2 NP_039228.1 1 107
XM_011527911.1 XP_011526213.1 2 64
GenBank mRNA BC130360.1 - 1 107
BC132969.1 - 1 107 40146991

View Full Product Details