Gene Transcripts

Gene Symbol
TANC2
Entrez Gene ID
26115
Gene Name
tetratricopeptide repeat, ankyrin repeat and coiled-coil containing 2
Gene Aliases
ROLSA, rols
Chromosome Location
Chr. 17: 62966523 - 63427706 on Build GRCh38
UniGene ID
Hs.410889
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025185.3 NP_079461.2 5 543
XM_005257203.4 XP_005257260.1 6 770
XM_006721806.3 XP_006721869.1 7 992
XM_006721807.3 XP_006721870.1 7 992
XM_006721810.3 XP_006721873.1 7 992
XM_006721811.3 XP_006721874.1 7 992
XM_011524597.2 XP_011522899.1 7 992
XM_011524598.2 XP_011522900.1 7 992
XM_011524599.1 XP_011522901.1 7 818
XM_011524600.2 XP_011522902.1 6 881
XM_011524601.2 XP_011522903.1 7 881
XM_011524602.2 XP_011522904.1 6 770
XM_017024429.1 XP_016879918.1 7 818
XM_017024430.1 XP_016879919.1 3 4159
XM_017024431.1 XP_016879920.1 7 992
XM_017024432.1 XP_016879921.1 7 992
GenBank mRNA AB046856.2 - 1 144

View Full Product Details