Gene Transcripts

Gene Symbol
HSF5
Entrez Gene ID
124535
Gene Name
heat shock transcription factor 5
Gene Aliases
HSF 5, HSTF 5
Chromosome Location
Chr. 17: 58420167 - 58488401 on Build GRCh38
UniGene ID
Hs.380061
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001080439.2 NP_001073908.2 6 2950
XM_011524283.1 XP_011522585.1 6 2963
GenBank mRNA AL137385.1 - 1 977
BC033017.1 - 6 2914 4825807
BC033020.1 - 6 2937 4823233
BC115385.1 - 7 2526 40038134
BC115386.1 - 7 2522

View Full Product Details