Gene Transcripts

Gene Symbol
ZFP92
Entrez Gene ID
139735
Gene Name
ZFP92 zinc finger protein
Gene Aliases
ZNF897
Chromosome Location
Chr. X: 153411461 - 153426481 on Build GRCh38
UniGene ID
Hs.534554
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001136273.1 NP_001129745.1 1, 2 41
XM_005274652.3 XP_005274709.1 3, 4 351
XM_011531115.2 XP_011529417.1 2, 3 302
XM_017029281.1 XP_016884770.1 4, 5 700

View Full Product Details