Gene Transcripts

Gene Symbol
IGSF23
Entrez Gene ID
147710
Gene Name
immunoglobulin superfamily member 23
Gene Aliases
Chromosome Location
Chr. 19: 44612577 - 44636781 on Build GRCh38
UniGene ID
Hs.512509
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001205280.1 NP_001192209.1 5 822
XM_017026319.1 XP_016881808.1 4 931
XM_017026323.1 XP_016881812.1 5 996
XM_017026324.1 XP_016881813.1 3 777
XM_017026325.1 XP_016881814.1 4 842
GenBank mRNA BG197674.1 - 2 265

View Full Product Details