Gene Transcripts

Gene Symbol
IGSF9B
Entrez Gene ID
22997
Gene Name
immunoglobulin superfamily member 9B
Gene Aliases
Chromosome Location
Chr. 11: 133908564 - 133956960 on Build GRCh38
UniGene ID
Hs.204121
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001277285.1 NP_001264214.1 14 2008
XM_006718795.3 XP_006718858.1 3 522
XM_011542691.2 XP_011540993.1 15 2695
XM_011542692.2 XP_011540994.1 15 2683
XM_011542694.2 XP_011540996.1 15 2698
XM_011542695.2 XP_011540997.1 14 2226
XM_017017393.1 XP_016872882.1 14 2713
XM_017017394.1 XP_016872883.1 15 2698
XM_017017395.1 XP_016872884.1 15 2697
XM_017017396.1 XP_016872885.1 8 2037
GenBank mRNA AB028953.1 - 2 250
AK097578.1 - 11 1738

View Full Product Details