Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001080437.1 | NP_001073906.1 | 8 | 1262 | |
XM_011510931.2 | XP_011509233.1 | 9 | 1576 | ||
XM_011510932.2 | XP_011509234.1 | 9 | 1578 | ||
XM_011510933.2 | XP_011509235.1 | 9 | 1582 | ||
XM_011510934.2 | XP_011509236.1 | 9 | 1582 | ||
XM_011510937.2 | XP_011509239.1 | 9 | 1581 | ||
XM_017003773.1 | XP_016859262.1 | 9 | 1578 | ||
XM_017003774.1 | XP_016859263.1 | 8 | 1347 | ||
XM_017003775.1 | XP_016859264.1 | 9 | 1580 | ||
XM_017003776.1 | XP_016859265.1 | 9 | 1578 | ||
XM_017003778.1 | XP_016859267.1 | 8 | 1342 | ||
XM_017003779.1 | XP_016859268.1 | 9 | 1278 | ||
XM_017003780.1 | XP_016859269.1 | 8 | 1134 | ||
XM_017003781.1 | XP_016859270.1 | 8 | 1124 | ||
XM_017003782.1 | XP_016859271.1 | 4 | 473 | ||
GenBank mRNA | AF439717.1 | - | 2 | 165 | |
AK074062.1 | - | 7 | 870 |