Gene Transcripts
- Gene Symbol
- TUSC7
- Entrez Gene ID
- 285194
- Gene Name
- tumor suppressor candidate 7 (non-protein coding)
- Gene Aliases
- LINC00902, LSAMP-AS1, LSAMP-AS3, LSAMPAS3, NCRNA00295
- Chromosome Location
- Chr. 3: 116709788 - 116717040 on Build GRCh38
- UniGene ID
- Hs.120364
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_015391.1 |
|
4 |
1847 |
|
GenBank mRNA |
BC035185.2 |
- |
4 |
1505 |
|
View Full Product Details