Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001040011.1 | NP_001035100.1 | 3 | 509 | |
NM_001318089.1 | NP_001305018.1 | 3 | 389 | ||
NM_001318092.1 | NP_001305021.1 | 3 | 250 | ||
XM_011518652.2 | XP_011516954.1 | 3 | 524 | ||
GenBank mRNA | AK311408.1 | - | 3 | 414 | |
BC029911.1 | - | 3 | 433 | ||
BG541668.1 | - | 3 | 292 | ||
CF129530.1 | - | 3 | 250 | ||
DQ414820.1 | - | 3 | 509 |