Gene Transcripts
- Gene Symbol
- TBC1D19
- Entrez Gene ID
- 55296
- Gene Name
- TBC1 domain family member 19
- Gene Aliases
- Chromosome Location
- Chr. 4: 26583924 - 26858919 on Build GRCh38
- UniGene ID
- Hs.479403
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_018317.3 |
NP_060787.2
|
2, 3 |
450 |
|
XM_011513852.2 |
XP_011512154.1
|
2, 3 |
290 |
|
XM_017008363.1 |
XP_016863852.1
|
2, 3 |
290 |
|
XM_017008364.1 |
XP_016863853.1
|
2, 3 |
290 |
|
GenBank mRNA |
AK001944.1 |
- |
2, 3 |
290 |
|
AK225395.1 |
- |
2, 3 |
290 |
|
AK289631.1 |
- |
2, 3 |
290 |
|
AK310795.1 |
- |
2, 3 |
297 |
|
BC031642.1 |
- |
2, 3 |
246 |
5175317 |
DC299740.1 |
- |
2, 3 |
450 |
|
View Full Product Details