Gene Transcripts

Gene Symbol
WDR12
Entrez Gene ID
55759
Gene Name
WD repeat domain 12
Gene Aliases
YTM1
Chromosome Location
Chr. 2: 202876958 - 202912226 on Build GRCh38
UniGene ID
Hs.73291
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018256.3 NP_060726.3 4, 5 1091
GenBank mRNA AF242546.1 - 4, 5 474
AK001743.1 - 4, 5 528
AK022781.1 - 4, 5 536
AK022782.1 - 4, 5 536
AK056092.1 - 4, 5 1091
BC008082.2 - 4, 5 471 3050219
EF011617.1 - 4, 5 470

View Full Product Details