Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001143962.1 | NP_001137434.1 | 2 | 347 | |
XM_017001267.1 | XP_016856756.1 | 2 | 347 | ||
XM_017001268.1 | XP_016856757.1 | 2 | 347 | ||
GenBank mRNA | BC157893.1 | - | 2 | 284 | |
DA874912.1 | - | 2 | 347 |