Gene Transcripts

Gene Symbol
BLOC1S4
Entrez Gene ID
55330
Gene Name
biogenesis of lysosomal organelles complex 1 subunit 4
Gene Aliases
BCAS4L, BLOS4, CNO
Chromosome Location
Chr. 4: 6716115 - 6717660 on Build GRCh38
UniGene ID
Hs.7570
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018366.2 NP_060836.1 1 1348
GenBank mRNA AK002092.1 - 1 1348

View Full Product Details