Gene Transcripts

Gene Symbol
BRK1
Entrez Gene ID
55845
Gene Name
BRICK1, SCAR/WAVE actin nucleating complex subunit
Gene Aliases
C3orf10, HSPC300, MDS027, hHBrk1
Chromosome Location
Chr. 3: 10115649 - 10127190 on Build GRCh38
UniGene ID
Hs.649307
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018462.4 NP_060932.2 3 272
GenBank mRNA AY148219.1 - 3 272
AY148220.1 - 3 272
BC001067.1 - 3 248 2823162
BC007929.1 - 3 241 3688264
BC019303.1 - 3 239 2823162
BC061515.1 - 3 246

View Full Product Details