Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001962.2 | NP_001953.1 | 5 | 2071 | |
XM_006714565.2 | XP_006714628.1 | 4 | 3459 | ||
XM_011543250.2 | XP_011541552.1 | 5 | 1884 | ||
XM_011543251.2 | XP_011541553.1 | 5 | 3397 | ||
XM_017009205.1 | XP_016864694.1 | 5 | 3160 |