Gene Transcripts
- Gene Symbol
- CDRT15L2
- Entrez Gene ID
- 256223
- Gene Name
- CMT1A duplicated region transcript 15-like 2
- Gene Aliases
- Chromosome Location
- Chr. 17: 20579724 - 20580911 on Build GRCh38
- UniGene ID
- Hs.131916
- Species
- Homo sapiens
- Gene Symbol
- CDRT15P2
- Entrez Gene ID
- 644694
- Gene Name
- CMT1A duplicated region transcript 15 pseudogene 2
- Gene Aliases
- CDRT15L1
- Chromosome Location
- Chr. 17: 15764703 - 15765689 on Build GRCh38
- UniGene ID
- Hs.513966
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_033865.1 |
|
3 |
433 |
|
View Full Product Details