Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001080471.1 | NP_001073940.1 | 21 | 2790 | |
XM_005245141.3 | XP_005245198.1 | 22 | 2983 | ||
XM_011509510.2 | XP_011507812.1 | 22 | 2922 | ||
XM_011509511.2 | XP_011507813.1 | 21 | 2878 | ||
XM_011509512.2 | XP_011507814.1 | 21 | 2802 | ||
XM_017001234.1 | XP_016856723.1 | 21 | 4048 | ||
XM_017001235.1 | XP_016856724.1 | 21 | 3918 | ||
XM_017001236.1 | XP_016856725.1 | 21 | 2885 | ||
XM_017001237.1 | XP_016856726.1 | 22 | 3129 | ||
XM_017001238.1 | XP_016856727.1 | 22 | 3068 | ||
XM_017001239.1 | XP_016856728.1 | 22 | 2879 | ||
XM_017001240.1 | XP_016856729.1 | 19 | 2447 | ||
XM_017001241.1 | XP_016856730.1 | 19 | 2595 | ||
XM_017001242.1 | XP_016856731.1 | 19 | 2499 | ||
GenBank mRNA | AK074121.1 | - | 16 | 2402 |