Gene Transcripts
- Gene Symbol
- SH3TC1
- Entrez Gene ID
- 54436
- Gene Name
- SH3 domain and tetratricopeptide repeats 1
- Gene Aliases
- Chromosome Location
- Chr. 4: 8182072 - 8241103 on Build GRCh38
- UniGene ID
- Hs.479116
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001318480.1 |
NP_001305409.1
|
6, 7 |
909 |
|
NM_018986.4 |
NP_061859.3
|
6, 7 |
826 |
|
XM_006713889.2 |
XP_006713952.1
|
3, 4 |
244 |
|
XM_011513488.1 |
XP_011511790.1
|
6, 7 |
803 |
|
XM_017008300.1 |
XP_016863789.1
|
6, 7 |
935 |
|
XM_017008301.1 |
XP_016863790.1
|
6, 7 |
699 |
|
XM_017008302.1 |
XP_016863791.1
|
3, 4 |
244 |
|
GenBank mRNA |
AK000363.1 |
- |
2, 3 |
214 |
|
AK074402.1 |
- |
6, 7 |
703 |
|
AK126165.1 |
- |
6, 7 |
782 |
|
AK222865.1 |
- |
2, 3 |
214 |
|
View Full Product Details