Gene Transcripts

Gene Symbol
SNTG1
Entrez Gene ID
54212
Gene Name
syntrophin gamma 1
Gene Aliases
G1SYN, SYN4
Chromosome Location
Chr. 8: 49909726 - 50796656 on Build GRCh38
UniGene ID
Hs.584914
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001287813.2 NP_001274742.1 5 565
NM_001287814.2 NP_001274743.1 3 717
NM_001321773.1 NP_001308702.1 3 717
NM_001321775.1 NP_001308704.1 5 565
NM_001321776.1 NP_001308705.1 4 792
NM_001321777.1 NP_001308706.1 3 717
NM_001321778.1 NP_001308707.1 4 523
NM_018967.4 NP_061840.1 4 792
NR_135794.1 4 792
NR_135795.1 4 792
NR_135796.1 4 792
NR_135797.1 4 792
NR_135798.1 3 717
XM_017013579.1 XP_016869068.1 6 3721
XM_017013580.1 XP_016869069.1 4 315
XM_017013581.1 XP_016869070.1 4 1037
XM_017013582.1 XP_016869071.1 4 1037
GenBank mRNA AJ003030.1 - 3 353
AK314084.1 - 5 502
AL161971.1 - 4 687
BC075072.2 - 2 157 30915592
BC104829.1 - 3 239 8143832
BC143695.1 - 3 239

View Full Product Details