Gene Transcripts
- Gene Symbol
- DDX12P
- Entrez Gene ID
- 440081
- Gene Name
- DEAD/H-box helicase 12, pseudogene
- Gene Aliases
- CHLR2, DDX12
- Chromosome Location
- Chr. 12: 9417691 - 9448172 on Build GRCh38
- UniGene ID
- Hs.447869
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_033399.1 |
|
13, 14 |
1617 |
|
GenBank mRNA |
BC065494.1 |
- |
4, 5 |
217 |
|
U33834.1 |
- |
10, 11 |
978 |
|
View Full Product Details