Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_018990.3 | NP_061863.1 | 2 | 322 | |
XM_006724763.1 | XP_006724826.1 | 2 | 289 | ||
GenBank mRNA | AK292023.1 | - | 2 | 270 | |
AK296665.1 | - | 2 | 224 | ||
AK314869.1 | - | 2 | 289 | ||
AL049683.1 | - | 2 | 247 | ||
BC051881.1 | - | 2 | 254 | 6011968 | |
CR975883.1 | - | 2 | 322 |